missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorobutyl)-1-cyclohexyltetrazole, TRC

Product Code. 30367360
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30367360 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367360 Supplier Toronto Research Chemicals Supplier No. TRCC36503810G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(4-Chlorobutyl)-1-cyclohexyltetrazole
CAS 73963-42-5
Molecular Formula C11 H19 Cl N4
Quantity 10 g
Synonym 1H-Tetrazole, 5-(4-chlorobutyl)-1-cyclohexyl-, 5-(4-Chlorobutyl)-1-cyclohexyl-1H-tetrazole, 1-Cyclohexyl-5-(4-chlorobutyl)-1,2,3,4-tetrazole, 1-Cyclohexyl-5-(4-chlorobutyl)tetrazole
InChI Formula InChI=1S/C11H19ClN4/c12-9-5-4-8-11-13-14-15-16(11)10-6-2-1-3-7-10/h10H,1-9H2
SMILES ClCCCCc1nnnn1C2CCCCC2
IUPAC Name 5-(4-chlorobutyl)-1-cyclohexyltetrazole
Molecular Weight (g/mol) 242.75
Formula Weight 242.1298
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.