missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-[(4-Fluorobenzyl)amino]-1H-imidazo[4,5-b]pyridin-2(3H)-one, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
5-[(4-Fluorobenzyl)amino]-1H-imidazo[4,5-b]pyridin-2(3H)-one
Specifications
Specifications
| Chemical Name or Material | 5-[(4-Fluorobenzyl)amino]-1H-imidazo[4,5-b]pyridin-2(3H)-one |
| CAS | 951624-49-0 |
| Analyte or Component Names | 5-[(4-Fluorobenzyl)amino]-1H-imidazo[4,5-b]pyridin-2(3H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H11 F N4 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-[[(4-Fluorophenyl)methyl]amino]-1,3-dihydro-2H-Imidazo[4,5-b]pyridin-2-one |
| InChI Formula | InChI=1S/C13H11FN4O/c14-9-3-1-8(2-4-9)7-15-11-6-5-10-12(17-11)18-13(19)16-10/h1-6H,7H2,(H3,15,16,17,18,19) |
| SMILES | Fc1ccc(CNc2ccc3NC(=O)Nc3n2)cc1 |
| IUPAC Name | 5-[(4-fluorophenyl)methylamino]-1,3-dihydroimidazo[4,5-b]pyridin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction