Learn More
5,5-Dimethyl-1,3-dioxane, TRC
Specifications
Specifications
| Chemical Name or Material | 5,5-Dimethyl-1,3-dioxane |
| CAS | 872-98-0 |
| Molecular Formula | C6 H12 O2 |
| Quantity | 500 mg |
| Synonym | 5,5-Dimethyl-m-dioxane, NSC 139437 |
| InChI Formula | InChI=1S/C6H12O2/c1-6(2)3-7-5-8-4-6/h3-5H2,1-2H3 |
| SMILES | CC1(C)COCOC1 |
| IUPAC Name | 5,5-dimethyl-1,3-dioxane |
| Molecular Weight (g/mol) | 116.158 |
| Formula Weight | 116.0837 |