Learn More
Invitrogen™ 5(6)-FAM, SE (5-(and-6)-Carboxyfluorescein, Succinimidyl Ester), mixed isomers
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | 5(6)-FAM SE |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | FITC (Fluorescein) |
| Label Type | Classic Dyes |