missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress 5(6)-TAMRA SE
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 527.53 g/mol |
| Solubility | DMSO : 10 mg/mL (18.96 mM; ultrasonic and warming and heat to 60°C) |
| Content And Storage | -20°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 246256-50-8 |
| Synonym | 5(6)-Carboxytetramethylrhodamine N-succinimidyl ester |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction