missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamido-2-nitrobenzoic acid, 98%, Thermo Scientific™

Product Code. 11428957
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11428957

Brand: Thermo Scientific Alfa Aesar B20568.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4368-83-6
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
MDL Number MFCD00035889
InChI Key ZSHFMOUMOUOGKI-UHFFFAOYSA-N
Synonym 5-acetamido-2-nitrobenzoic acid, 5-acetylamino-2-nitrobenzoic acid, benzoic acid, 5-acetylamino-2-nitro, 2-nitro-5-acetylaminobenzoic acid, 5-acetamido-2-nitrobenzoicacid, pubchem13220, acmc-209jv7, 5-acetamido-2-nitrobenzoic
PubChem CID 78076
ChEBI CHEBI:86569
SMILES CC(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Quantity 50 g
Formula Weight 224.17
Percent Purity 98%
Chemical Name or Material 5-Acetamido-2-nitrobenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.