Learn More
5-Amino-2-hydroxybenzonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-hydroxybenzonitrile |
| CAS | 87029-84-3 |
| Molecular Formula | C7 H6 N2 O |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H6N2O/c8-4-5-3-6(9)1-2-7(5)10/h1-3,10H,9H2 |
| SMILES | Nc1ccc(O)c(c1)C#N |
| IUPAC Name | 5-amino-2-hydroxybenzonitrile |
| Molecular Weight (g/mol) | 134.135 |
| Formula Weight | 134.048 |