Learn More
5-Amino-2-methylbenzotrifluoride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-methylbenzotrifluoride |
| CAS | 65934-74-9 |
| Molecular Formula | C8H8F3N |
| Quantity | 2.5 g |
| Synonym | (4-Methyl-3-trifluoromethylphenyl)amine, 3-Trifluoromethyl-4-methylaniline, 4-Amino-2-trifluoromethyltoluene, 4-Methyl-3-(trifluoromethyl)aniline, 4-Methyl-3-(trifluoromethyl)benzenamine |
| InChI Formula | InChI=1S/C8H8F3N/c1-5-2-3-6(12)4-7(5)8(9,10)11/h2-4H,12H2,1H3 |
| SMILES | Cc1ccc(N)cc1C(F)(F)F |
| IUPAC Name | 4-methyl-3-(trifluoromethyl)aniline |
| Molecular Weight (g/mol) | 175.15 |
| Formula Weight | 175.0609 |