Learn More
5-Aminovaleric Acid Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Aminovaleric Acid Hydrochloride |
| CAS | 627-95-2 |
| Molecular Formula | C5H11NO2 . HCl |
| Quantity | 10 g |
| Synonym | 5-Aminopentanoic Acid Hydrochloride, 5-Aminovaleric Acid Hydrochloride, delta-Aminovaleric Acid Hydrochloride |
| InChI Formula | InChI=1S/C5H11NO2.ClH/c6-4-2-1-3-5(7)8;/h1-4,6H2,(H,7,8);1H |
| SMILES | Cl.NCCCCC(=O)O |
| IUPAC Name | 5-aminopentanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 153.61 |
| Formula Weight | 153.0557 |