Learn More
Edmund Buehler™ 5° Angle Tilt Shaker
Rich, robust, reliable tilt shaker WS 10 that works with slow, linear tilting motion and a fix angle of tilt of 5°.
Brand: Edmund Buehler™ 6114 000
Additional Details : Weight : 5.00000kg
Description
- Tilt Shaker WS 10 works with slow, linear tilting motion and a fix angle of tilt of 5°
- Shaking speed of 1 - 23 rpm
- Shaking platform of 340 x 340 mm can be loaded with a weight of 10 kg
- Basic device includes basic tray and rubber mat
Spécification
5° | |
1 to 23 rpm | |
230 V | |
50/60 Hz | |
205 mm | |
WS 10 | |
Basic tray and Rubber mat |
10 kg | |
230 V, 50/60 Hz | |
5°C to 50°C | |
450 mm | |
355 mm | |
85% | |
1 to 120 min. |