Learn More
5% Biodiesel/95% High Cetane Diesel Fuel, v/v, VHG™
Description
5% Biodiesel/95% High Cetane Diesel Fuel, v/v
Specifications
Specifications
| Chemical Name or Material | Bio Diesel Fuel |
| CAS | 870530-78-2 |
| Analyte or Component Names | Bio Diesel Fuel |
| Shipping Condition | Room Temperature |
| Concentration | 5% |
| Synonym | Diesel fuel substitutes, biodiesel, B 10, B 10 biofuel, B 100, B 100 biofuel, B 20, B 20 biofuel, B 5, B 5 biofuel, B 50, B 50 biofuel, Biodiesel, Biofuels, biodiesel, Biodiesel fuel |
| IUPAC Name | Biodiesel Fuel |
| Solvent or Matrix | High Cetane Diesel Fuel/Biodiesel |
| Quantity | 20 mL |