Learn More
5-Bromo-2-chloropyridine, 98%, Thermo Scientific™
Catalog Number p-3777614
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
53939-30-3
C5H3BrClN
192.44
MFCD01318951
PEAOEIWYQVXZMB-UHFFFAOYSA-N
2-chloro-5-bromopyridine, 5-bromo-2-chloro-pyridine, pyridine, 5-bromo-2-chloro, 2-chloro-5-bromo-pyridine, 2-chlorine-5-bromine pyridine, pubchem1168, acmc-1afhf, 3-bromo-6-chloropyridine, 2-chloro-5-bromo pyridine, 3-bromo-6-chloro-pyridine
2734414
5-bromo-2-chloropyridine
C1=CC(=NC=C1Br)Cl
53939-30-3
C5H3BrClN
192.44
MFCD01318951
PEAOEIWYQVXZMB-UHFFFAOYSA-N
2-chloro-5-bromopyridine, 5-bromo-2-chloro-pyridine, pyridine, 5-bromo-2-chloro, 2-chloro-5-bromo-pyridine, 2-chlorine-5-bromine pyridine, pubchem1168, acmc-1afhf, 3-bromo-6-chloropyridine, 2-chloro-5-bromo pyridine, 3-bromo-6-chloro-pyridine
2734414
5-bromo-2-chloropyridine
C1=CC(=NC=C1Br)Cl
CAS Min % 97.5
CAS Max % 100.0
Melting Point 67°C to 71°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 192.44
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5-Bromo-2-chloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-chloropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.