missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzotrifluoride, 97%, Thermo Scientific™

Catalog Number p-7077289
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
393-37-3
C7H3BrF4
243.00
MFCD00042493
AJLIJYGWAXPEOK-UHFFFAOYSA-N
5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
2736325
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
FC1=CC=C(Br)C=C1C(F)(F)F
This item is not returnable. View return policy
393-37-3
C7H3BrF4
243.00
MFCD00042493
AJLIJYGWAXPEOK-UHFFFAOYSA-N
5-bromo-2-fluorobenzotrifluoride, 4-bromo-1-fluoro-2-trifluoromethyl benzene, 2-fluoro-5-bromobenzotrifluoride, 4-fluoro-3-trifluoromethyl bromobenzene, 5-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 5-bromo-2-fluoro-trifluoromethyl benzene, 2-fluoro-5-bromo benzotrifluoride, 1-bromo-4-fluoro-3-trifluoromethyl benzene, benzene, 4-bromo-1-fluoro-2-trifluoromethyl, acmc-209j4s
2736325
4-bromo-1-fluoro-2-(trifluoromethyl)benzene
FC1=CC=C(Br)C=C1C(F)(F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 142.0°C to 147.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-fluorobenzotrifluoride
compliance-icons
Product Identifier
  • 5-BROMO-2-FLUOROBENZOTRIFLUORIDE, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'