missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxyaniline, 97%

Product Code. p-7028400
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11481717

Brand: Thermo Scientific Alfa Aesar B25693.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6358-77-6 | C7H8BrNO | 202.05 g/mol

5-Bromo-2-methoxyaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-methoxyaniline is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Preserve in well- closed, light- resistant & tight container. Store in cool & dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6358-77-6
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.05
MDL Number MFCD04037882
InChI Key OPGNSNDTPPIYPG-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy aniline, 2-amino-4-bromoanisole, 5-bromo-o-anisidine, benzenamine, 5-bromo-2-methoxy, 5-bromo-2-methoxy-aniline, 4-bromo-2-aminoanisole, 2-methoxy-5-bromoaniline, acmc-1b9k2, 5-bromo-2-methoxyphenylamine, 5-bromo-2-methyloxy aniline
PubChem CID 3585328
IUPAC Name 5-bromo-2-methoxyaniline
SMILES COC1=C(N)C=C(Br)C=C1

Specifications

Melting Point 94°C to 96°C
Quantity 5 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 202.05
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxyaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.