Learn More
5-Bromo-2-methylphenol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 36138-76-8 |
| Molecular Formula | C7H7BrO |
| MDL Number | MFCD08061906 |
| Quantity | 25 g |
| SMILES | BrC1=CC(=C(C=C1)C)O |
| Molecular Weight (g/mol) | 187.03 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Bromo-2-methylphenol |