Learn More
5-Bromo-N,N-Dimethyltryptamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Bromo-N,N-Dimethyltryptamine Hydrochloride |
| CAS | 2613384-12-4 |
| Molecular Formula | C12H15N2Br • (HCl) |
| Quantity | 100 mg |
| Synonym | NSC 622272, [2-(5-Bromo-1H-indol-3-yl)ethyl]dimethylamine Hydrochloride,5-Bromo-N,N-dimethyl-1H-indole-3-ethanamine Hydrochloride,2-(5-Bromo-1H-indol-3-yl)-N,N-dimethylethanamine Hydrochloride |
| InChI Formula | InChI=1S/C12H15BrN2.ClH/c1-15(2)6-5-9-8-14-12-4-3-10(13)7-11(9)12;/h3-4,7-8,14H,5-6H2,1-2H3;1H |
| SMILES | BrC1=CC2=C(NC=C2CCN(C)C)C=C1.Cl |
| IUPAC Name | 2-(5-bromo-1H-indol-3-yl)-N,N-dimethylethan-1-amine hydrochloride |
| Molecular Weight (g/mol) | 303.63 |
| Formula Weight | 302.02 |