Learn More
5-Bromo-pyridine-2-sulfonyl chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Bromo-pyridine-2-sulfonyl chloride |
| CAS | 874959-68-9 |
| Molecular Formula | C5H3BrClNO2S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C5H3BrClNO2S/c6-4-1-2-5(8-3-4)11(7,9)10/h1-3H |
| SMILES | ClS(=O)(=O)c1ccc(Br)cn1 |
| IUPAC Name | 5-bromopyridine-2-sulfonyl chloride |
| Molecular Weight (g/mol) | 256.5 |
| Formula Weight | 254.8756 |