missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopicolinic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4323378
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
30766-11-1
C6H3BrNO2
201.00
MFCD00234149
MNNQIBXLAHVDDL-UHFFFAOYSA-M
5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
608544
[O-]C(=O)C1=CC=C(Br)C=N1
This item is not returnable. View return policy
30766-11-1
C6H3BrNO2
201.00
MFCD00234149
MNNQIBXLAHVDDL-UHFFFAOYSA-M
5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
608544
[O-]C(=O)C1=CC=C(Br)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 177.0°C to 178.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 5-bromopyridine-2-carboxylic acid
Formula Weight 202.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Bromopicolinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromopicolinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'