missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyrimidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7055455
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
37131-87-6
C5H3BrN2O2
202.995
MFCD00496793
XGPTXUYKEDPXCO-UHFFFAOYSA-N
5-bromo-pyrimidine-2-carboxylic acid, 5-bromo-2-pyrimidinecarboxylic acid, 2-pyrimidinecarboxylic acid, 5-bromo, 5-bromopyrimidine-2-carboxylicacid, pubchem16922, acmc-1agpr, ksc222c2t, 5-bromopyrimidine-2-caboxylic acid, abbypharma ap-10-1676, 2-pyrimidinecarboxylicacid, 5-bromo
16462970
5-bromopyrimidine-2-carboxylic acid
C1=C(C=NC(=N1)C(=O)O)Br
This item is not returnable. View return policy
37131-87-6
C5H3BrN2O2
202.995
MFCD00496793
XGPTXUYKEDPXCO-UHFFFAOYSA-N
5-bromo-pyrimidine-2-carboxylic acid, 5-bromo-2-pyrimidinecarboxylic acid, 2-pyrimidinecarboxylic acid, 5-bromo, 5-bromopyrimidine-2-carboxylicacid, pubchem16922, acmc-1agpr, ksc222c2t, 5-bromopyrimidine-2-caboxylic acid, abbypharma ap-10-1676, 2-pyrimidinecarboxylicacid, 5-bromo
16462970
5-bromopyrimidine-2-carboxylic acid
C1=C(C=NC(=N1)C(=O)O)Br

5-Bromopyrimidine-2-carboxylic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyrimidine-2-carboxylic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 193°C to 198°C
Quantity 250 mg
Beilstein 121432
Solubility Information Slightly soluble in water.
Formula Weight 203
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromopyrimidine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'