Learn More
5-Chloro-1-Indanone, TRC
Product Code. 30361649
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361649 5 g -
30361797 25 g 25g
30361179 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30361649 Supplier Toronto Research Chemicals Supplier No. TRCC3672355G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42348-86-7
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.6
IUPAC Name 5-chloro-2,3-dihydroinden-1-one
SMILES Clc1ccc2C(=O)CCc2c1
Specifications
Quantity 5 g
Synonym 5-Chloro- 1-indanone, 5-Chloro-1-indanone, 5-Chloro-2,3-dihydro-1H-inden-1-one, 5-Chloro-2,3-dihydroinden-1-one
InChI Formula InChI=1S/C9H7ClO/c10-7-2-3-8-6(5-7)1-4-9(8)11/h2-3,5H,1,4H2
Formula Weight 166.0185
Chemical Name or Material 5-Chloro-1-Indanone