missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%

Product Code. p-7047308
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369435 10 g 10g
11379435 100 g 100g
11389435 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369435 Supplier Thermo Scientific Alfa Aesar Supplier No. L18655.09
Add to basket
This item is not returnable. View return policy

CAS: 3380-34-5 | C12H7Cl3O2 | 289.536 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
5-Chloro-2-(2,4-dichlorophenoxy)phenol, acts as a bacteriostat and preservative for cosmetic and detergent compositions. It is also used as an antiseptic, disinfectant and anticonvulsant. It is one of the main ingredient in shampoos, deodorants, toothpastes, mouth washes and household cleaning products. It inhibits the growth of bacteria, fungi on the surfaces of the textile, thereby it imparts odor protection to wool, synthetics, blends and non-wovens.

Solubility
Soluble in ethanol, methanol, diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.

Notes
Store in cool place. Decomposes on heating. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3380-34-5
Molecular Formula C12H7Cl3O2
Molecular Weight (g/mol) 289.536
MDL Number MFCD00800992
InChI Key XEFQLINVKFYRCS-UHFFFAOYSA-N
Synonym triclosan, 5-chloro-2-2,4-dichlorophenoxy phenol, irgasan, 2,4,4'-trichloro-2'-hydroxydiphenyl ether, cloxifenolum, triclosanum, irgasan dp300, stri-dex face wash, aquasept, sapoderm
PubChem CID 5564
ChEBI CHEBI:164200
IUPAC Name 5-chloro-2-(2,4-dichlorophenoxy)phenol
SMILES C1=CC(=C(C=C1Cl)O)OC2=C(C=C(C=C2)Cl)Cl

Specifications

Melting Point 56°C to 59°C
Quantity 10 g
UN Number UN3077
Beilstein 2057142
Merck Index 14,9657
Solubility Information Soluble in ethanol,methanol,diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.
Formula Weight 289.55
Percent Purity 99%
Chemical Name or Material 5-Chloro-2-(2,4-dichlorophenoxy)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.