Learn More
5-Chloro-2,4-dinitrobenzoic acid, 98%
Product Code. 11467156
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467156 5 g 5g
11477156 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11467156 Supplier Thermo Scientific Alfa Aesar Supplier No. B25479.06
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
CAS: 136833-36-8 | C7H3ClN2O6 | 246.559 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 136833-36-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.559
MDL Number MFCD00091829
InChI Key VVQOEJCXWMGEJG-UHFFFAOYSA-N
Synonym 5-chloro-2,4-dinitrobenzoic acid, benzoic acid, 5-chloro-2,4-dinitro, acmc-20a30o, 2,4-dinitro-5-chlorobenzoic acid, 3-chloro-4,6-dinitrobenzoic acid
PubChem CID 3727860
SMILES C1=C(C(=CC(=C1Cl)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
Specifications
Melting Point 180°C to 182°C
Quantity 5 g
Formula Weight 246.56
Percent Purity 98%
Chemical Name or Material 5-Chloro-2,4-dinitrobenzoic acid