missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-sulfamoylbenzoic acid, 97%

Product Code. 11420118
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11420118

Brand: Thermo Scientific Alfa Aesar B23698.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22117-85-7 | C8H9NO5S | 231.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22117-85-7
Molecular Formula C8H9NO5S
Molecular Weight (g/mol) 231.222
MDL Number MFCD00129997
InChI Key SQAILWDRVDGLGY-UHFFFAOYSA-N
Synonym 2-methoxy-5-aminosulfonylbenzoic acid, benzoic acid, 5-aminosulfonyl-2-methoxy, 5-sulphamoyl-o-anisic acid, 2-methoxy-5-sulfonylbenzoic acid, sulpiride impurity d, acmc-209fsc, 2-methoxy-5-sulfamoylbenzoicacid, 2-methoxy-5-sulphamoylbenzoic acid, 2-methoxy-5-sulfamoyl benzoic acid, 2-methoxy-5-sulfamoyl-benzoic acid
PubChem CID 89601
IUPAC Name 2-methoxy-5-sulfamoylbenzoic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)N)C(=O)O

Specifications

Quantity 250 g
Formula Weight 232.23
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-sulfamoylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.