Learn More
Tocris Bioscience™ (±)-5'-Chloro-5'-deoxy-ENBA
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
(±)-5'-Chloro-5'-deoxy-ENBA is a highly selective adenosine A1 receptor agonist (Ki values are 0.51, 1290, 1340 and 2740 nM at A1, A3, A2A and A2B receptors respectively). Reverses formalin-induced nocifensive behavior in mice; antinociceptive.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | (±)-5'-Chloro-5'-deoxy-ENBA |
| Purity | >98% |
| Molecular Weight (g/mol) | 379.84 |