Learn More
5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine, Mikromol™
Product Code. 30049906
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30049906
Brand: Mikromol MM0521.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine
CAS 1784008-01-0
Analyte or Component Names 5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine
Shipping Condition Room Temperature
Molecular Formula C9 H14 Cl N5
Certifications/Compliance ISO 17025
Synonym 5-Chloro-6-(1-piperidinyl)-2,4-pyrimidinediamine
InChI Formula InChI=1S/C9H14ClN5/c10-6-7(11)13-9(12)14-8(6)15-4-2-1-3-5-15/h1-5H2,(H4,11,12,13,14)
SMILES Nc1nc(N)c(Cl)c(n1)N2CCCCC2
IUPAC Name 5-chloro-6-piperidin-1-ylpyrimidine-2,4-diamine
Molecular Weight (g/mol) 227.69
Quantity 25 mg
Formula Weight 227.0938 g/mol