missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Chlorobenzofuroxan, 97%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Melting Point | 45°C to 48°C |
| Quantity | 5 g |
| Formula Weight | 170.56 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Chlorobenzofuroxan |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction