Learn More
5-(Difluoromethoxy)-3-fluorobenzyl bromide, JRD, 97%, Thermo Scientific™
Product Code. 15458255
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15458255
Brand: Thermo Scientific Alfa Aesar H32969.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017779-39-3
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.034
MDL Number MFCD09832362
InChI Key SZFFWIBFEJPELW-UHFFFAOYSA-N
Synonym 1-bromomethyl-3-difluoromethoxy-5-fluorobenzene, 5-difluoromethoxy-3-fluorobenzyl bromide, jrd, 5-difluoromethoxy-3-fluorobenzyl bromide, 5-bromomethyl-3-fluorophenoxy difluoromethane
PubChem CID 46737678
IUPAC Name 1-(bromomethyl)-3-(difluoromethoxy)-5-fluorobenzene
SMILES C1=C(C=C(C=C1OC(F)F)F)CBr
Specifications
Quantity 250 mg
UN Number UN3265
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 5-(Difluoromethoxy)-3-fluorobenzyl bromide, JRD