Learn More
5-Ethyl-2-fluorophenylboronic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Ethyl-2-fluorophenylboronic acid |
| CAS | 900175-03-3 |
| Molecular Formula | C8H10BFO2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H10BFO2/c1-2-6-3-4-8(10)7(5-6)9(11)12/h3-5,11-12H,2H2,1H3 |
| SMILES | CCc1ccc(F)c(c1)B(O)O |
| Molecular Weight (g/mol) | 167.97 |
| Formula Weight | 168.0758 |