Learn More
5-Ethyl-2-methoxybenzene-1-sulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Ethyl-2-methoxybenzene-1-sulfonyl Chloride |
| CAS | 88040-88-4 |
| Molecular Formula | C9H11O3SCl |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H11ClO3S/c1-3-7-4-5-8(13-2)9(6-7)14(10,11)12/h4-6H,3H2,1-2H3 |
| SMILES | CCc1ccc(OC)c(c1)S(=O)(=O)Cl |
| IUPAC Name | 5-ethyl-2-methoxybenzenesulfonyl chloride |
| Molecular Weight (g/mol) | 234.69 |
| Formula Weight | 234.0117 |