Learn More
5-Ethyl-2-methoxybenzene-1-sulfonyl Chloride, TRC
Product Code. 30370565
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370565 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30370565 Supplier Toronto Research Chemicals Supplier No. TRCE93723550MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Ethyl-2-methoxybenzene-1-sulfonyl Chloride
CAS 88040-88-4
Molecular Formula C9H11O3SCl
Quantity 50 mg
InChI Formula InChI=1S/C9H11ClO3S/c1-3-7-4-5-8(13-2)9(6-7)14(10,11)12/h4-6H,3H2,1-2H3
SMILES CCc1ccc(OC)c(c1)S(=O)(=O)Cl
IUPAC Name 5-ethyl-2-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol) 234.69
Formula Weight 234.0117