missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoronicotinic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Melting Point | 193°C to 198°C |
Color | White |
Quantity | 25 g |
Solubility Information | Soluble in methanol. |
Formula Weight | 141.1 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 5-Fluoronicotinic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'