Learn More
Invitrogen™ 5-Formylcytosine (5-fC) Recombinant Rabbit Monoclonal Antibody (RM477)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA544549
Product Code. 17826731
  • € 452.00 / 50µg
Description
This antibody reacts to dTyr alpha Tubulin.
5-formylcytosine is an oxidized derivative of 5-methylcytosine, a type of DNA methylation. 5-formylcytosine is a nucleobase analogue with a cytosine that possesses a hydrogen at position 5 that has been replaced by a formyl group. 5-methylcytosine is oxidized to 5-hydroxymethylcytosine which is then oxidized to 5-formylcytosine and later to 5-carboxylcytosine. These oxidations are catalyzed by the Ten-Eleven Translocation (TET) enzyme family. 5-formylcytosine has been shown to determine nucleosome organization and plays a role in regulating transcription.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5-Formylcytosine (5-fC)
Recombinant Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
KLH-conjugated 5-Formylcytosines.
50 μg
Primary
-20°C
Liquid
Chemical
Dot Blot, ELISA
RM477
Unconjugated
Rabbit
Protein A
RUO
Chemical