missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-hmC Antibody (513), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43568
This item is not returnable.
View return policy
Description
5-hmC Monoclonal antibody specifically detects 5-hmC in Nonspecies specific samples. It is validated for Immunoprecipitation, Dot Blot, Methylated DNA Immunoprecipitation
Specifications
| 5-hmC | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Immunoprecipitation, Dot Blot, Methylated DNA Immunoprecipitation | |
| 513 | |
| Immunoprecipitation, Dot Blot 1:100 - 1:2000, Methylated DNA Immunoprecipitation | |
| 5'-hmC, 5-hydroxymethylcytosine | |
| The immunogen used to generate this antibody corresponds to 5-Hydroxymethylcytosine/ 5-hmC | |
| 0.1 mL | |
| DNA Repair, Epigenetics, Stem Cells | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction