missing translation for 'onlineSavingsMsg'
Learn More

5-hmC Antibody (AB3/63.3), FITC, Novus Biologicals™

Product Code. 18666126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18666126 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18666126 Supplier Novus Biologicals Supplier No. NBP250099F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

5-hmC Monoclonal antibody specifically detects 5-hmC in All samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-hmC
Applications Immunofluorescence
Classification Monoclonal
Clone AB3/63.3
Conjugate FITC
Formulation PBS
Gene Alias 5'-hmC, 5-hydroxymethylcytosine
Host Species Rat
Immunogen 5-Hydroxymethylcytosine conjugated to BSA
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Stem Cells
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.