missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-hmC Antibody (AB3/63.3), Janelia Fluor™ 549, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50099JF549
This item is not returnable.
View return policy
Description
5-hmC Monoclonal antibody specifically detects 5-hmC in All samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dot Blot
Specifications
| 5-hmC | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 5'-hmC, 5-hydroxymethylcytosine | |
| 5-hmC conjugated to BSA | |
| 0.1 mL | |
| DNA Repair, Epigenetics, Stem Cells | |
| All species | |
| Purified |
| Immunofluorescence, Immunoprecipitation, Dot Blot | |
| AB3/63.3 | |
| 50 mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction