Learn More
5-HT2A Antibody (1055220), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB112501X
Product Code. 30063543
  • € 619.00 / 0.10mL
Description
5-HT2A Monoclonal antibody specifically detects 5-HT2A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5-HT2A
Monoclonal
Alexa Fluor 532
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Immunocytochemistry
1055220
Flow Cytometry, Immunocytochemistry
5-HT-2, 5-HT2 receptor, 5-HT-2A, 5-hydroxytryptamine (serotonin) receptor 2A, 5-hydroxytryptamine receptor 2A, HTR2,5-HT2A, serotonin 5-HT-2A receptor, Serotonin receptor 2A
Synthetic peptide corresponding to the amino acids 1-75 in the N-term extracellular domain of the Serotonin Receptor 5-HT2A (HTR2A)., Accession # P28223
0.1 mL
GPCR
3356
Store at 4C in the dark.