missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT2A Antibody (1055220), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB112501D
This item is not returnable.
View return policy
Description
5-HT2A Monoclonal antibody specifically detects 5-HT2A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| 5-HT2A | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 1055220 | |
| Flow Cytometry, Immunocytochemistry | |
| 5-HT-2, 5-HT2 receptor, 5-HT-2A, 5-hydroxytryptamine (serotonin) receptor 2A, 5-hydroxytryptamine receptor 2A, HTR2,5-HT2A, serotonin 5-HT-2A receptor, Serotonin receptor 2A | |
| Synthetic peptide corresponding to the amino acids 1-75 in the N-term extracellular domain of the Serotonin Receptor 5-HT2A (HTR2A)., Accession # P28223 | |
| 0.1 mL | |
| GPCR | |
| 3356 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction