missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5L2/GPR77 Antibody (468705), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4729G
This item is not returnable.
View return policy
Description
C5L2/GPR77 Monoclonal antibody specifically detects C5L2/GPR77 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| C5L2/GPR77 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27202 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 468705 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| C5a anaphylatoxin chemotactic receptor C5L2, C5L2G-protein coupled receptor 77, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77 | |
| Chinese hamster ovary cell line transfected with mouse C5L2, Met1-Val344, Accession # Q8BW93 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction