Learn More
5-HT2B Antibody (CL14576) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26870MFV610
Product Code. 30295265
  • € 836.00 / 0.10mL
Description
5-HT2B Monoclonal antibody specifically detects 5-HT2B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5-HT2B
Monoclonal
mFluor Violet 610 SE
5-HT(2B), 5-HT-2B, 5-HT2B5-HT 2B receptor, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, Serotonin receptor 2B
This antibody was developed using a recombinant protein derived from P41595, with the exact immunogen sequence remaining proprietary.
0.1 mL
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
3357
Store at 4⪚C in the dark.
IgG1
Western Blot
CL14576
50mM Sodium Borate
Mouse
Protein A purified