missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT2C Antibody (496214) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5764B
This item is not returnable.
View return policy
Description
5-HT2C Monoclonal antibody specifically detects 5-HT2C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| 5-HT2C | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 496214 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| 5-HT1C, 5-HT-1C, 5-HT-2C, 5-HT2C5-hydroxytryptamine receptor 1C, 5-HTR2C, 5-hydroxytryptamine (serotonin) receptor 2C, 5-hydroxytryptamine receptor 2C, HTR1C, serotonin 5-HT-2C receptor, Serotonin receptor 2C | |
| NS0 mouse myeloma cell line transfected with human 5HT2C | |
| 0.1 mL | |
| GPCR | |
| 3358 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction