missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT3B Antibody (4G6H7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61792-0.025ml
This item is not returnable.
View return policy
Description
5-HT3B Monoclonal specifically detects 5-HT3B in Human samples. It is validated for Western Blot, ELISA.
Specifications
| 5-HT3B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| 5-HT3-B, 5-HT3B5-hydroxytryptamine receptor 3B, 5-hydroxytryptamine (serotonin) receptor 3B, 5-hydroxytryptamine 3 receptor B subunit, Serotonin receptor 3B, serotonin-gated ion channel subunit | |
| Mouse | |
| 50.3 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 4G6H7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HTR3B | |
| Purified recombinant fragment of human HTR3B (AA: extra 23-150) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 9177 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction