missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78403DL594
This item is not returnable.
View return policy
Description
5-HT4 Polyclonal antibody specifically detects 5-HT4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| 5-HT4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 594 | |
| 5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, Serotonin receptor 4 | |
| A synthetic peptide made to a C-terminal portion of the human 5HT4 Receptor protein (between residues 350-388) [UniProt Q13639, isoform 1] | |
| 0.1 mL | |
| Asthma, GPCR, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3360 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction