missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT7 Antibody (1015322) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10325K
This item is not returnable.
View return policy
Description
5-HT7 Monoclonal antibody specifically detects 5-HT7 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| 5-HT7 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 1015322 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
| Synthetic peptide containing human 5-HTR7, Accession # P34969 | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3363 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction