missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT7 Antibody (SR1954), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22298-100ul
This item is not returnable.
View return policy
Description
5-HT7 Monoclonal antibody specifically detects 5-HT7 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| 5-HT7 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1954 | |
| Western Blot 1:500-1:2000 | |
| 5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
| A synthesized peptide derived from human 5-HT7 (Uniprot #: P34969) | |
| 100 μg | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3363 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction