Learn More
5-Hydroxy-1,4-naphthoquinone, 99%
Product Code. 11472010
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472010 5 g 5g
11462010 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11472010 Supplier Thermo Scientific Alfa Aesar Supplier No. H28343.06
 Restricted Item
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
CAS: 481-39-0 | C10H6O3 | 174.16 g/mol
5-Hydroxy-1,4-naphthoquinone is used as a natural dye in cloth, fabrics, wool and ink. It is a coloring agent for food and cosmetics. It is involved in the synthesis of poly(hydroxyl-1,4-naphthoquinone) stabilized gold nanoparticles (AuNQ NPs), which is used for nonenzymatic electrochemical detection of glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Hydroxy-1,4-naphthoquinone is used as a natural dye in cloth, fabrics, wool and ink. It is a coloring agent for food and cosmetics. It is involved in the synthesis of poly(hydroxyl-1,4-naphthoquinone) stabilized gold nanoparticles (AuNQ NPs), which is used for nonenzymatic electrochemical detection of glucose.
Solubility
Soluble in alcohol, chloroform, dimethylsulfoxide, ether and aqueous solution of alkalies. Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong bases, strong reducing agents and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 481-39-0
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.16
MDL Number MFCD00001684
InChI Key KQPYUDDGWXQXHS-UHFFFAOYSA-N
Synonym juglone, 5-hydroxy-1,4-naphthoquinone, 5-hydroxy-1,4-naphthalenedione, regianin, walnut extract, juglon, nucin, 5-hydroxynaphthoquinone, akhnot, yuglon
PubChem CID 3806
ChEBI CHEBI:15794
IUPAC Name 5-hydroxynaphthalene-1,4-dione
SMILES OC1=CC=CC2=C1C(=O)C=CC2=O
Specifications
Melting Point 161°C to 163°C
Quantity 5 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,5629
Solubility Information Soluble in alcohol,chloroform,dimethylsulfoxide,ether and aqueous solution of alkalies. Slightly soluble in water.
Formula Weight 174.16
Percent Purity 99%
Chemical Name or Material 5-Hydroxy-1,4-naphthoquinone