missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-N-methylprotriptyline, TRC

Product Code. 30011909
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011909 10 mg 10mg
30138514 100 mg 100mg
30005725 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011909 Supplier Toronto Research Chemicals Supplier No. TRCH94803510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18029-54-4
Formula Weight 293.178
InChI Formula InChI=1S/C20H23NO/c1-21(2)15-7-14-20(22)18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)20/h3-6,8-13,22H,7,14-15H2,1-2H3
IUPAC Name 11-[3-(dimethylamino)propyl]dibenzo[2,1-b:1',2'-e][7]annulen-11-ol
Molecular Formula C20 H23 N O
Molecular Weight (g/mol) 293.4
SMILES CN(C)CCCC1(O)c2ccccc2C=Cc3ccccc13
Synonym 5H-Dibenzo[a,d]cyclohepten-5-ol, 5-[3-(dimethylamino)propyl]-
Chemical Name or Material 5-[3-(Dimethylamino)propyl]-5H-dibenzo[a,d]cyclohepten-5-ol

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.