Learn More
5-Indanol, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Indanol |
| CAS | 1470-94-6 |
| Molecular Formula | C9H10O |
| Quantity | 5 g |
| Synonym | 2,3-Dihydro-1H-inden-5-ol, 2,3-Dihydroinden-5-ol, 5-Hydroxyhydrindene, 5-Hydroxyindan, 5-Hydroxyindane, Indan-6-ol, NSC 9775 |
| InChI Formula | InChI=1S/C9H10O/c10-9-5-4-7-2-1-3-8(7)6-9/h4-6,10H,1-3H2 |
| SMILES | Oc1ccc2CCCc2c1 |
| Molecular Weight (g/mol) | 134.18 |
| Formula Weight | 134.0732 |