missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Lipoxygenase, p Ser523 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-277
This item is not returnable.
View return policy
Description
5-Lipoxygenase Polyclonal specifically detects 5-Lipoxygenase in Human, Rat samples. It is validated for Western Blot.
Specifications
| 5-Lipoxygenase (p Ser523) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| P09917 | |
| ALOX5 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser523 conjugated to KLH. Accession # P09917 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼80k 5-LO phosphorylated at Ser523 in Western blots. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| 5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204 | |
| Rabbit | |
| 80 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience, Phospho Specific | |
| 240 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction