Learn More
5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole, Mikromol™
Product Code. 17953746
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953746 25 mg 25mg
17982276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17953746 Supplier Mikromol Supplier No. MM0095.240025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole
CAS 2555002-22-5
Analyte or Component Names 5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H17 N3 O2
Certifications/Compliance ISO 17025
Synonym 5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole
InChI Formula InChI=1S/C16H17N3O2/c1-9-8-17-14(10(2)15(9)21-4)16-18-12-6-5-11(20-3)7-13(12)19-16/h5-8H,1-4H3,(H,18,19)
SMILES COc1ccc2[nH]c(nc2c1)c3ncc(C)c(OC)c3C
IUPAC Name 5-methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole
Molecular Weight (g/mol) 283.33
Quantity 25 mg
Formula Weight 283.1321 g/mol