Learn More
5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzo[d]imidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole |
| CAS | 2555002-22-5 |
| Molecular Formula | C16 H17 N3 O2 |
| Quantity | 250 mg |
| Synonym | 5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole, 1H-Benzimidazole, 6-methoxy-2-(4-methoxy-3,5-dimethyl-2-pyridinyl)- (ACI), 6-Methoxy-2-(4-methoxy-3,5-dimethyl-2-pyridinyl)-1H-benzimidazole (ACI) |
| InChI Formula | InChI=1S/C16H17N3O2/c1-9-8-17-14(10(2)15(9)21-4)16-18-12-6-5-11(20-3)7-13(12)19-16/h5-8H,1-4H3,(H,18,19) |
| SMILES | COc1ccc2[nH]c(nc2c1)c3ncc(C)c(OC)c3C |
| IUPAC Name | 5-methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole |
| Molecular Weight (g/mol) | 283.33 |
| Formula Weight | 283.1321 |